Skip to Content
Merck
F.A.M.E. Mix RM-4 certified reference material, ampule of 100 mg (Neat)

F.A.M.E. Mix RM-4

F.A.M.E. Mix RM-4 certified reference material, ampule of 100 mg (Neat)
Synonym(s):
AOCS Reference Mixtures, Oil Reference Standard, AOCS

Products

Product Category
Brand
Format
Functional Group
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing