Ethyl ethanesulfonate
Synonym(s):
Ethanesulfonic acid ethyl ester
Empirical Formula (Hill Notation):
C4H10O3S
CAS Number:
Molecular Weight:
138.19
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing