Skip to Content
Merck
Ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate 98%

Ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate

Ethyl 6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate 98%
Synonym(s):
3-Ethoxycarbonyl-6,7,8-trifluoro-1,4-hydro-4-oxoquinoline
Empirical Formula (Hill Notation):
C12H8F3NO3
CAS Number:
Molecular Weight:
271.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing