Skip to Content
Merck
Ethyl 2-bromocaprylate ≥97.0% (GC)

Ethyl 2-bromocaprylate

Ethyl 2-bromocaprylate ≥97.0% (GC)
Synonym(s):
Ethyl 2-bromooctanoate
Linear Formula:
CH3(CH2)5CHBrCOOC2H5
CAS Number:
Molecular Weight:
251.16
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing