Skip to Content
Merck
Ethanol – 2-propanol mixture BioUltra, for molecular biology, 19:1, A15 ISO1, ≥99.5% (GC)

Ethanol – 2-propanol mixture

Ethanol – 2-propanol mixture BioUltra, for molecular biology, 19:1, A15 ISO1, ≥99.5% (GC)
Synonym(s):
Ethanol : 2-propanol, Ethyl alcohol:Isopropyl alcohol
Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07

Products

Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Showing 1-1 of 1