Ethametsulfuron-methyl
Synonym(s):
Methyl 2-[(4-ethoxy-6-methylamino-1,3,5-triazin-2-yl)carbamoylsulfamoyl]benzoate, Methyl 2-[[[[[4-ethoxy-6-(methylamino)-1,3,5-triazin-2-yl]amino]carbonyl]amino]sulfonyl]benzoate
Empirical Formula (Hill Notation):
C15H18N6O6S
CAS Number:
Molecular Weight:
410.41
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing