Skip to Content
Merck
Epiamastatin hydrochloride ≥97% (HPLC)

Epiamastatin hydrochloride

Epiamastatin hydrochloride ≥97% (HPLC)
Synonym(s):
[(2R,3R)-3-Amino-2-hydroxy-5-methylhexanoyl]-Val-Val-Asp hydrochloride
Empirical Formula (Hill Notation):
C21H38N4O8
CAS Number:
Molecular Weight:
474.55

Products

Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing