
D-myo-Inositol 1,4-bisphosphate potassium salt

Empirical Formula (Hill Notation):
C6H12K2O12P2
CAS Number:
Molecular Weight:
416.30
Products
Product Category
Brand
Biological Source
Formula Weight
Purity
Tissue
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing