Skip to Content
Merck
D-Mannose 6-phosphate barium salt hydrate ≥98%

D-Mannose 6-phosphate barium salt hydrate

D-Mannose 6-phosphate barium salt hydrate ≥98%
Empirical Formula (Hill Notation):
C6H11BaO9P · xH2O
Molecular Weight:
395.45 (anhydrous basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing