<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
VN
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
DL-Thiorphan
Synonym(s):
(±)-N-(3-Mercapto-2-benzylpropionyl)glycine, DL-3-Mercapto-2-benzylpropanoylglycine, DL-Thiorphan
Empirical Formula (Hill Notation):
C
12
H
15
NO
3
S
CAS Number:
76721-89-6
Molecular Weight:
253.32
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T6031
DL-Thiorphan,
≥98% (TLC)
Expand
View Pricing
598510
DL-Thiorphan,
A thiol containing amido-acid that selectively binds to the active site zinc of metalloproteinases and blocks their activity (IC₅₀ = 2.1 nM for neutral endopeptidase-NEP).
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Tiếng Việt - VI
English - EN
Learn More