Skip to Content
Merck
DL-tert-Leucine 98%

DL-tert-Leucine

DL-tert-Leucine 98%
Synonym(s):
(±)-2-Amino-3,3-dimethylbutyric acid, DL-α-tert-Butylglycine
Linear Formula:
(CH3)3CCH(NH2)COOH
CAS Number:
Molecular Weight:
131.17

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing