Skip to Content
Merck
DL-p-Chloroamphetamine hydrochloride ≥94% (HPLC)

DL-p-Chloroamphetamine hydrochloride

DL-p-Chloroamphetamine hydrochloride ≥94% (HPLC)
Synonym(s):
4-Chloro-α-methylphenethylamine hydrochloride, PCA
Linear Formula:
ClC6H4CH2CH(CH3)NH2·HCl
CAS Number:
Molecular Weight:
206.11

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing