
DL-Amphetamine hydrochloride

Empirical Formula (Hill Notation):
C9H13N · HCl
CAS Number:
Molecular Weight:
171.67
Products
Brand
Formula Weight
Purity
Format
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing