![(±)-α-Methyl-(4-carboxyphenyl)glycine](/deepweb/assets/sigmaaldrich/product/structures/325/522/a8299575-03ea-456c-b1c7-6b408a0c152f/640/a8299575-03ea-456c-b1c7-6b408a0c152f.png)
(±)-α-Methyl-(4-carboxyphenyl)glycine
![(±)-α-Methyl-(4-carboxyphenyl)glycine](/deepweb/assets/sigmaaldrich/product/structures/325/522/a8299575-03ea-456c-b1c7-6b408a0c152f/640/a8299575-03ea-456c-b1c7-6b408a0c152f.png)
Synonym(s):
(±)-MCPG
Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20
Products
Product Category
Brand
Formula Weight
Gene Alias
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing