Skip to Content
Merck
DL-Alanine ethyl ester hydrochloride ≥99.0% (AT)

DL-Alanine ethyl ester hydrochloride

DL-Alanine ethyl ester hydrochloride ≥99.0% (AT)
Linear Formula:
CH3CH(NH2)COOC2H5 · HCl
CAS Number:
Molecular Weight:
153.61
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing