
Dirhodium tetracaprolactamate

Synonym(s):
Tetrakis[μ-(hexahydro-2H-azepin-2-onato-κN1,κO2)]dirhodium
Empirical Formula (Hill Notation):
C24H40N4O4Rh2
CAS Number:
Molecular Weight:
654.41
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing