Skip to Content
Merck
(−)-Dimethyl D-tartrate 99%

(−)-Dimethyl D-tartrate

(−)-Dimethyl D-tartrate 99%
Synonym(s):
D-(−)-Tartaric acid dimethyl ester, Dimethyl D-tartrate
Linear Formula:
[-CH(OH)CO2CH3]2
CAS Number:
Molecular Weight:
178.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing