Skip to Content
Merck
Dimethyl acetylsuccinate

Dimethyl acetylsuccinate

Dimethyl acetylsuccinate
Synonym(s):
2-Acetylbutanedioic acid-1,4-dimethyl ester
Linear Formula:
CH3O2CCH2CH(COCH3)CO2CH3
CAS Number:
Molecular Weight:
188.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing