Skip to Content
Merck
Dimethyl 5-bromoisophthalate ≥98.0% (GC)

Dimethyl 5-bromoisophthalate

Dimethyl 5-bromoisophthalate ≥98.0% (GC)
Synonym(s):
Dimethyl 5-bromobenzene-1,3-dicarboxylate
Empirical Formula (Hill Notation):
C10H9BrO4
CAS Number:
Molecular Weight:
273.08
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing