Skip to Content
Merck
Dimethyl 2-oxoheptylphosphonate technical grade, 85%

Dimethyl 2-oxoheptylphosphonate

Dimethyl 2-oxoheptylphosphonate technical grade, 85%
Synonym(s):
(2-Oxoheptyl)phosphonic acid dimethyl ester, Dimethyl (hexanoylmethyl)phosphonate
Linear Formula:
CH3(CH2)4COCH2P(O)(OCH3)2
CAS Number:
Molecular Weight:
222.22
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing