![(Diethylamino)sulfur trifluoride solution 1.0 M in dichloromethane](/deepweb/assets/sigmaaldrich/product/structures/336/810/a7d674d0-ab88-4c01-b2ab-973c5ce37ae7/640/a7d674d0-ab88-4c01-b2ab-973c5ce37ae7.png)
(Diethylamino)sulfur trifluoride solution
![(Diethylamino)sulfur trifluoride solution 1.0 M in dichloromethane](/deepweb/assets/sigmaaldrich/product/structures/336/810/a7d674d0-ab88-4c01-b2ab-973c5ce37ae7/640/a7d674d0-ab88-4c01-b2ab-973c5ce37ae7.png)
Empirical Formula (Hill Notation):
C4H10F3NS
CAS Number:
Molecular Weight:
161.19
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing