Skip to Content
Merck
Dichloromethylphosphine 96%

Dichloromethylphosphine

Dichloromethylphosphine 96%
Synonym(s):
Methyldichlorophosphine
Linear Formula:
CH3PCl2
CAS Number:
Molecular Weight:
116.91
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing