Skip to Content
Merck
Dibenzyl dicarbonate 92%

Dibenzyl dicarbonate

Dibenzyl dicarbonate 92%
Synonym(s):
Dibenzyl pyrocarbonate, Z2O
Linear Formula:
(CO2CH2C6H5)2O
CAS Number:
Molecular Weight:
286.28

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing