Skip to Content
Merck
(DHQD)2AQN 95%

(DHQD)2AQN

(DHQD)2AQN 95%
Synonym(s):
Hydroquinidine (anthraquinone-1,4-diyl) diether
Empirical Formula (Hill Notation):
C54H56N4O6
CAS Number:
Molecular Weight:
857.05

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing