Skip to Content
Merck
DH-FTTF sublimed, 95%

DH-FTTF

DH-FTTF sublimed, 95%
Synonym(s):
5,5′-Bis(7-hexyl-9H-fluoren-2-yl)-2,2′-bithiophene
Empirical Formula (Hill Notation):
C46H46S2
CAS Number:
Molecular Weight:
662.99

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing