D-Allylglycine
Synonym(s):
(R)-(+)-2-Amino-4-pentenoic acid, D-2-Amino-4-pentenoic acid
Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing