Skip to Content
Merck
(+)-6-Aminopenicillanic acid analytical standard

(+)-6-Aminopenicillanic acid

(+)-6-Aminopenicillanic acid analytical standard
Synonym(s):
(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-APA
Empirical Formula (Hill Notation):
C8H12N2O3S
CAS Number:
Molecular Weight:
216.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing