Skip to Content
Merck
(+)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid 97%

(+)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid

(+)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid 97%
Synonym(s):
(2R,3R,11R,12R)-1,4,7,10,13,16-Hexaoxacyclooctadecane-2,3,11,12-tetracarboxylic acid
Empirical Formula (Hill Notation):
C16H24O14
CAS Number:
Molecular Weight:
440.35

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing