Skip to Content
Merck
Cyclanilide PESTANAL®, analytical standard

Cyclanilide

Cyclanilide PESTANAL®, analytical standard
Synonym(s):
1-[(2,4-Dichlorophenyl)aminocarbonyl]-1-cyclopropanecarboxylic acid
Empirical Formula (Hill Notation):
C11H9Cl2NO3
CAS Number:
Molecular Weight:
274.10
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing