![C-Undecylcalix[4]resorcinarene monohydrate 99%](/deepweb/assets/sigmaaldrich/product/structures/413/929/c4d12c06-2c4c-473d-a7c2-9baab1d8e314/640/c4d12c06-2c4c-473d-a7c2-9baab1d8e314.png)
C-Undecylcalix[4]resorcinarene monohydrate
![C-Undecylcalix[4]resorcinarene monohydrate 99%](/deepweb/assets/sigmaaldrich/product/structures/413/929/c4d12c06-2c4c-473d-a7c2-9baab1d8e314/640/c4d12c06-2c4c-473d-a7c2-9baab1d8e314.png)
Empirical Formula (Hill Notation):
C72H112O8 · H2O
CAS Number:
Molecular Weight:
1123.67
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing