Skip to Content
Merck
Copper(I) thiophenolate ≥98.0% (RT)

Copper(I) thiophenolate

Copper(I) thiophenolate ≥98.0% (RT)
Synonym(s):
Copper(I) thiophenoxide, Phenylthiocopper(I), Thiophenol copper(I) salt
Linear Formula:
C6H5SCu
CAS Number:
Molecular Weight:
172.71

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing