Colby trifluoromethylation reagent
Synonym(s):
Hexafluoroacetone hydrate-1,8-diazo-bicyclo[5.4.0]undec-7-ene salt
Empirical Formula (Hill Notation):
C12H18F6N2O2
CAS Number:
Molecular Weight:
336.27
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing