Skip to Content
Merck
Clostridium perfringens Vitroids 200 CFU 200 CFU, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Clostridium perfringens Vitroids 200 CFU

Clostridium perfringens Vitroids 200 CFU 200 CFU, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Clostridium perfringens Vitroids

Products

Brand
Manufacturer
Physical Form
Application
Microorganism Suitability
Special Grade