Skip to Content
Merck
Clopyralid (2-hydroxyethyl)ammonium PESTANAL®, analytical standard

Clopyralid (2-hydroxyethyl)ammonium

Clopyralid (2-hydroxyethyl)ammonium PESTANAL®, analytical standard
Synonym(s):
3,6-Dichloropyridine-2-carboxylic acid ethanolammonium salt
Empirical Formula (Hill Notation):
C8H10Cl2N2O3
CAS Number:
Molecular Weight:
253.08
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing