Skip to Content
MilliporeSigma
(Chloromethyl)-isopropoxy-dimethylsilane 97%

(Chloromethyl)-isopropoxy-dimethylsilane

(Chloromethyl)-isopropoxy-dimethylsilane 97%
Synonym(s):
(Chloromethyl)dimethylisopropoxysilane
Linear Formula:
ClCH2Si(CH3)2OCH(CH3)2
CAS Number:
Molecular Weight:
166.72

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing