Skip to Content
Merck
Chloro-dimethyl-octadecylsilane solution produced by Wacker Chemie AG, Burghausen, Germany, 70% in toluene

Chloro-dimethyl-octadecylsilane solution

Chloro-dimethyl-octadecylsilane solution produced by Wacker Chemie AG, Burghausen, Germany, 70% in toluene
Synonym(s):
Dimethyl-octadecylchlorosilane, Octadecyldimethylchlorosilane
Empirical Formula (Hill Notation):
C20H43ClSi
CAS Number:
Molecular Weight:
347.09

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group