Skip to Content
Merck
Chloro-decyl-dimethylsilane ≥97.0% (GC)

Chloro-decyl-dimethylsilane

Chloro-decyl-dimethylsilane ≥97.0% (GC)
Synonym(s):
Decyl-dimethyl-chlorosilane
Empirical Formula (Hill Notation):
C12H27ClSi
CAS Number:
Molecular Weight:
234.88
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing