
Chitosan from crab shells

Synonym(s):
2-Amino-2-deoxy-(1→4)-β-D-glucopyranan, Poly-(1,4-β-D-glucopyranosamine)
CAS Number:
Products
Product Category
Brand
Color
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing