Skip to Content
Merck
Chirald® 99%

Chirald®

Chirald® 99%
Synonym(s):
(2S,3R)-(+)-4-Dimethylamino-3-methyl-1,2-diphenyl-2-butanol, (2S,3R)-(+)-4-Dimethylamino-1,2-diphenyl-3-methyl-2-butanol, Dextrocarbinol base
Linear Formula:
C6H5CH2C(C6H5)(OH)CH(CH3)CH2N(CH3)2
CAS Number:
Molecular Weight:
283.41
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing