
Carprofen for system suitability

Synonym(s):
Carprofen, 6-Chloro-α-methyl-9H-carbazole-2-acetic acid
Empirical Formula (Hill Notation):
C15H12ClNO2
CAS Number:
Molecular Weight:
273.71
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing