
Butyldimethyl(dimethylamino)silane

Linear Formula:
CH3(CH2)3Si(CH3)2N(CH3)2
CAS Number:
Molecular Weight:
159.34
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing