Skip to Content
Merck
Butyl 4-hydroxybenzoate-ring-13C6 solution 50 μg/mL in acetone, analytical standard

Butyl 4-hydroxybenzoate-ring-13C6 solution

Butyl 4-hydroxybenzoate-ring-13C6 solution 50 μg/mL in acetone, analytical standard
Synonym(s):
4-Hydroxybenzoic acid n-butyl ester-ring-13C6 solution, Butyl paraben-ring-13C6 solution
Empirical Formula (Hill Notation):
13C6C5H14O3
CAS Number:
Molecular Weight:
200.18
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique