Skip to Content
Merck
Bromopolystyrene extent of labeling: ~4 mmol/g Br loading

Bromopolystyrene

Bromopolystyrene extent of labeling: ~4 mmol/g Br loading
Synonym(s):
Copolymer of styrene and divinylbenzene, brominated, Polystyrene crosslinked with divinylbenzene, brominated
Linear Formula:
[CH2CH(C6H4Br)]n
CAS Number:

Products

Brand
Physical Form
Chain Type
Reaction Type
Showing 1-2 of 2