Bromfenvinphos-ethyl
Empirical Formula (Hill Notation):
C12H14BrCl2O4P
CAS Number:
Molecular Weight:
404.02
Products
Product Category
Brand
Formula Weight
Product Line
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing