Skip to Content
Merck
Boron tribromide dimethyl sulfide complex 97%

Boron tribromide dimethyl sulfide complex

Boron tribromide dimethyl sulfide complex 97%
Synonym(s):
Dimethyl sulfide-tribromoborane, Tribromoborane-methyl sulfide
Linear Formula:
BBr3 · S(CH3)2
CAS Number:
Molecular Weight:
312.66

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing