Skip to Content
Merck
Boc-Phe(4-NO2)-OH ≥99.0% (T)

Boc-Phe(4-NO2)-OH

Boc-Phe(4-NO2)-OH ≥99.0% (T)
Synonym(s):
Boc-4-nitro-L-phenylalanine
Empirical Formula (Hill Notation):
C14H18N2O6
CAS Number:
Molecular Weight:
310.30
EC Number:

Products

Brand
Formula Weight
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing