Skip to Content
Merck
Boc-Gly-OtBu 99%

Boc-Gly-OtBu

Boc-Gly-OtBu 99%
Synonym(s):
Boc-glycine tert-butyl ester
Linear Formula:
(CH3)3COC(O)NHCH2CO2C(CH3)3
CAS Number:
Molecular Weight:
231.29

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing