Skip to Content
Merck
Boc-D-phenylalaninol 98%

Boc-D-phenylalaninol

Boc-D-phenylalaninol 98%
Synonym(s):
(R)-(+)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, (R)-2-(Boc-amino)-3-phenyl-1-propanol, N-(tert-Butoxycarbonyl)-D-phenylalaninol
Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
251.32

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing