Skip to Content
Merck
Boc-5-Ava-OH 97%

Boc-5-Ava-OH

Boc-5-Ava-OH 97%
Synonym(s):
5-(Boc-amino)pentanoic acid, 5-(Boc-amino)valeric acid
Linear Formula:
(CH3)3CO2CNH(CH2)4CO2H
CAS Number:
Molecular Weight:
217.26

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing