Skip to Content
Merck
Boc-4-phenyl-Phe-OH 97%

Boc-4-phenyl-Phe-OH

Boc-4-phenyl-Phe-OH 97%
Synonym(s):
(S)-2-(Boc-amino)-3-(4-biphenylyl)propionic acid, Boc-4-phenyl-L-phenylalanine, Boc-Bip-OH
Linear Formula:
C6H5C6H4CH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
341.40

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing